Please login to your online account to display your discounted pricing

2-Mercapto-4-methylpyrimidine hydrochloride, 99%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  125510250

 View more versions of this product

Catalog No. AC125510250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 162.64
Quantity 25g
Color Yellow
CAS Max % 100.0
Chemical Name or Material 2-Mercapto-4-methylpyrimidine hydrochloride
Assay Percent Range 99%
Molecular Formula C5H6N2S·HCl
MDL Number MFCD00012783
CAS 6959-66-6
CAS Min % 98.5
Infrared Spectrum Authentic
Synonym 4-Methyl-2-pyrimidinethiol hydrochloride

Specifications

Chemical Name or Material 2-Mercapto-4-methylpyrimidine hydrochloride
CAS 6959-66-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H6N2S·HCl
Formula Weight 162.64
MDL Number MFCD00012783
Synonym 4-Methyl-2-pyrimidinethiol hydrochloride