missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercapto-4-methylpyrimidine hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

CAS 6959-66-6
Molecular Formula C5H7ClN2S
Molecular Weight (g/mol) 162.635
MDL Number MFCD00012783
InChI Key UQJLPBLXSJWAKG-UHFFFAOYSA-N
Synonym 2-mercapto-4-methylpyrimidine hydrochloride, 4-methylpyrimidine-2-thiol hydrochloride, 4-methylpyrimidine-2 1h-thione hydrochloride, 6-methyl-1,2-dihydropyrimidine-2-thione hydrochloride, 4-methylpyrimidine-2-thiol hcl, 2 1h-pyrimidinethione, 4-methyl-, monohydrochloride, c5h6n2s.clh, 2-mercapto-4-methyl pyrimidine hydrochloride, pubchem21990, acmc-1b3ky
PubChem CID 2723697
IUPAC Name 6-methyl-1H-pyrimidine-2-thione;hydrochloride
SMILES CC1=CC=NC(=S)N1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC125511000 SDS View Documents Acros Organics
125511000
100g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC125515000 SDS View Documents Acros Organics
AC125515000
500g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

6959-66-6
162.635
UQJLPBLXSJWAKG-UHFFFAOYSA-N
2723697
CC1=CC=NC(=S)N1.Cl
C5H7ClN2S
MFCD00012783
2-mercapto-4-methylpyrimidine hydrochloride, 4-methylpyrimidine-2-thiol hydrochloride, 4-methylpyrimidine-2 1h-thione hydrochloride, 6-methyl-1,2-dihydropyrimidine-2-thione hydrochloride, 4-methylpyrimidine-2-thiol hcl, 2 1h-pyrimidinethione, 4-methyl-, monohydrochloride, c5h6n2s.clh, 2-mercapto-4-methyl pyrimidine hydrochloride, pubchem21990, acmc-1b3ky
6-methyl-1H-pyrimidine-2-thione;hydrochloride

Specifications

Authentic
Yellow
100g
98.5
99%
MFCD00012783
UQJLPBLXSJWAKG-UHFFFAOYSA-N
6-methyl-1H-pyrimidine-2-thione;hydrochloride
2723697
Fine Crystalline Powder
2-Mercapto-4-methylpyrimidine hydrochloride
Glass bottle
265.0°C
6959-66-6
100.0
C5H7ClN2S
2-mercapto-4-methylpyrimidine hydrochloride, 4-methylpyrimidine-2-thiol hydrochloride, 4-methylpyrimidine-2 1h-thione hydrochloride, 6-methyl-1,2-dihydropyrimidine-2-thione hydrochloride, 4-methylpyrimidine-2-thiol hcl, 2 1h-pyrimidinethione, 4-methyl-, monohydrochloride, c5h6n2s.clh, 2-mercapto-4-methyl pyrimidine hydrochloride, pubchem21990, acmc-1b3ky
CC1=CC=NC(=S)N1.Cl
162.635
162.64
99%