Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-4,6-dimethylpyrimidine sodium salt, 98%

$72.93 - $983.92

Chemical Identifiers

Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine sodium salt
CAS 41840-27-1
Molecular Formula C6H7N2NaS
Formula Weight 162.18
MDL Number MFCD04039775
Synonym 4,6-Dimethylpyrimidine-2-thiol sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2492509

  SDS
alfa aesar™
B2492509
10g Each for $72.93

AAB2492518

  SDS
alfa aesar™
B2492518
50g Each for $248.62

AAB2492530

  SDS
alfa aesar™
B2492530
250g Each for $983.92
Description & Specifications

Specifications

Formula Weight 162.18
Quantity 10g
Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine sodium salt
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C6H7N2NaS
MDL Number MFCD04039775
CAS 41840-27-1
Assay 98%
Synonym 4,6-Dimethylpyrimidine-2-thiol sodium salt
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.