Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-4,6-dimethylpyrimidine hydrate, 98%

$43.24 - $178.55

Chemical Identifiers

Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine hydrate
CAS 22325-27-5
Molecular Formula C6H8N2S·xH2O
Formula Weight 140.21 (Anhydrous)
MDL Number MFCD00006079
Beilstein 81330
Synonym 4,6-Dimethylpyrimidine-2-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1223109

 
alfa aesar™
A1223109
10g Each for $43.24

AAA1223118

 
alfa aesar™
A1223118
50g Each for $178.55
Description & Specifications

Specifications

Formula Weight 140.21 (Anhydrous)
Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine hydrate
Assay 98%
Molecular Formula C6H8N2S·xH2O
MDL Number MFCD00006079
CAS 22325-27-5
Beilstein 81330
Synonym 4,6-Dimethylpyrimidine-2-thiol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
RTECS Number UW5650000
Melting Point 213° to 216°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.