Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-4,5-diphenyloxazole, 98%

$37.05 - $37.05

Chemical Identifiers

Chemical Name or Material 2-Mercapto-4,5-diphenyloxazole
CAS 6670-13-9
Molecular Formula C15H11NOS
Formula Weight 253.33
MDL Number MFCD00173660
Synonym 4,5-Diphenyloxazole-2-thiol; 4,5-Diphenyl-2-mercaptooxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2011106

  SDS
alfa aesar™
B2011106
5g Each for $37.05
Description & Specifications

Specifications

Formula Weight 253.33
Health Hazard 1 Warning
CAS 6670-13-9
Melting Point 251° to 256°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Mercapto-4,5-diphenyloxazole
Molecular Formula C15H11NOS
Assay 98%
Synonym 4,5-Diphenyloxazole-2-thiol; 4,5-Diphenyl-2-mercaptooxazole
Quantity 5g
MDL Number MFCD00173660
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.