missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-4,5-dimethylthiazole, 95%

$29.45 - $345.43

Chemical Identifiers

CAS 5351-51-9
Molecular Formula C5H7NS2
Molecular Weight (g/mol) 145.238
MDL Number MFCD00037895
InChI Key KKHBRTFQIYIHEI-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazole-2-thiol, 4,5-dimethyl-thiazole-2-thiol, 4,5-dimethylthiazole-2 3h-thione, dimethyl-1,3-thiazole-2-thiol, 4,5-dimethyl-1,3-thiazole-2-thiol, 2 3h-thiazolethione, 4,5-dimethyl, 2-thiazolethiol, 4,5-dimethyl, 4,5-dimethyl-2-mercaptothiazole, 2-mercapto-4,5-dimethylthiazole, 4,5-dimethyl-2-thiazolethiol
PubChem CID 817026
IUPAC Name 4,5-dimethyl-3H-1,3-thiazole-2-thione
SMILES CC1=C(SC(=S)N1)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65085MD View Documents Alfa Aesar™
H65085MD
250mg Each for $29.45
 
AAH6508503 View Documents Alfa Aesar™
H6508503
1g Each for $86.99
 
AAH6508506 View Documents Alfa Aesar™
H6508506
5g Each for $345.43
 
Specifications

Chemical Identifiers

5351-51-9
145.238
KKHBRTFQIYIHEI-UHFFFAOYSA-N
817026
CC1=C(SC(=S)N1)C
C5H7NS2
MFCD00037895
4,5-dimethylthiazole-2-thiol, 4,5-dimethyl-thiazole-2-thiol, 4,5-dimethylthiazole-2 3h-thione, dimethyl-1,3-thiazole-2-thiol, 4,5-dimethyl-1,3-thiazole-2-thiol, 2 3h-thiazolethione, 4,5-dimethyl, 2-thiazolethiol, 4,5-dimethyl, 4,5-dimethyl-2-mercaptothiazole, 2-mercapto-4,5-dimethylthiazole, 4,5-dimethyl-2-thiazolethiol
4,5-dimethyl-3H-1,3-thiazole-2-thione

Specifications

250mg
C5H7NS2
107610
KKHBRTFQIYIHEI-UHFFFAOYSA-N
4,5-dimethyl-3H-1,3-thiazole-2-thione
817026
Solid
2-Mercapto-4,5-dimethylthiazole
5351-51-9
MFCD00037895
4,5-dimethylthiazole-2-thiol, 4,5-dimethyl-thiazole-2-thiol, 4,5-dimethylthiazole-2 3h-thione, dimethyl-1,3-thiazole-2-thiol, 4,5-dimethyl-1,3-thiazole-2-thiol, 2 3h-thiazolethione, 4,5-dimethyl, 2-thiazolethiol, 4,5-dimethyl, 4,5-dimethyl-2-mercaptothiazole, 2-mercapto-4,5-dimethylthiazole, 4,5-dimethyl-2-thiazolethiol
CC1=C(SC(=S)N1)C
145.238
145.24
95%