missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ketoglutaric acid, disodium salt, dihydrate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC271250010

 View more versions of this product

Catalog No. AC271250010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

200ppm max
500ppm max
12% to 16%
200ppm max
1kg
305-72-6
100.0
C5H4Na2O5·2H2O
MFCD00150702
2-ketoglutaric acid disodium salt, 2-oxoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium 2-oxoglutarate, disodium 2-oxopentanedioate, disodium ? ketoglutarate, disodium oxoglurate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, unii-flp7p4rm46
YBGBJYVHJTVUSL-UHFFFAOYSA-L
disodium;2-oxopentanedioate
31040
Crystalline Powder or Granules
2-Ketoglutaric acid, disodium salt, dihydrate
Authentic
Glass bottle
White
99%
98.5
99%
NaOOCCH2CH2C(O)COONa·2H2O
03, 1813
Solubility in water: soluble in water
C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]
226.1
226.1
99%

Chemical Identifiers

305-72-6
226.1
YBGBJYVHJTVUSL-UHFFFAOYSA-L
31040
C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]
C5H4Na2O5·2H2O
MFCD00150702
2-ketoglutaric acid disodium salt, 2-oxoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium 2-oxoglutarate, disodium 2-oxopentanedioate, disodium ? ketoglutarate, disodium oxoglurate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, unii-flp7p4rm46
disodium;2-oxopentanedioate
Safety and Handling

Safety and Handling

EINECSNumber : 206-167-8

TSCA : TSCA

SDS Documents