Learn More
2-Ketoglutaric acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1025636
Description
2-Ketoglutaric acid is used as an intermediate in the krebâs cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ketoglutaric acid is used as an intermediate in the krebâs cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.
Solubility
Soluble in water, methanol and dimethyl sulfoxide.
Notes
Store away from heat store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
2-Ketoglutaric acid | |
113°C to 117°C | |
MFCD00004165 | |
1705689 | |
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid | |
KPGXRSRHYNQIFN-UHFFFAOYSA-N | |
2-oxopentanedioic acid | |
51 | |
146.1 |
328-50-7 | |
C5H6O5 | |
500 g | |
14,5303 | |
Soluble in water,methanol and dimethyl sulfoxide. | |
C(CC(=O)O)C(=O)C(=O)O | |
146.098 | |
CHEBI:30915 | |
98% |
Chemical Identifiers
328-50-7 | |
146.098 | |
KPGXRSRHYNQIFN-UHFFFAOYSA-N | |
51 | |
2-oxopentanedioic acid |
C5H6O5 | |
MFCD00004165 | |
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid | |
CHEBI:30915 | |
C(CC(=O)O)C(=O)C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P305+P351+P338-P310
H318
EINECSNumber : 206-330-3
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only