missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ketoglutaric acid, 98%

Manufacturer:  Alfa Aesar™ A1025614

 View more versions of this product

Catalog No. AAA1025614



Description

Description

Applications
2-Ketoglutaric acid is used as an intermediate in the kreb’s cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.

Solubility
Soluble in water, methanol and dimethyl sulfoxide.

Notes
Store away from heat store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Ketoglutaric acid
25g
C5H6O5
1705689
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
C(CC(=O)O)C(=O)C(=O)O
146.098
CHEBI:30915
98%
113°C to 117°C
328-50-7
MFCD00004165
14,5303
KPGXRSRHYNQIFN-UHFFFAOYSA-N
2-oxopentanedioic acid
51
146.1

Chemical Identifiers

328-50-7
146.098
KPGXRSRHYNQIFN-UHFFFAOYSA-N
51
2-oxopentanedioic acid
C5H6O5
MFCD00004165
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
CHEBI:30915
C(CC(=O)O)C(=O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents