Please login to your online account to display your discounted pricing

2-Isopropylphenyl isothiocyanate, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Isopropylphenyl isothiocyanate
CAS 36176-31-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%+
Molecular Formula C10H11NS
Linear Formula CH3CH(CH3)C6H4NCS
Formula Weight 177.27
MDL Number MFCD00022053
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01626DA

 
fisher scientific company
SB01626DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.27
Quantity 1g
Density 1.0500g/mL
Assay 95%
CAS Min % 95.0
Linear Formula CH3CH(CH3)C6H4NCS
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 95%+
MDL Number MFCD00022053
CAS 36176-31-5
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material 2-Isopropylphenyl isothiocyanate
Molecular Formula C10H11NS
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.