missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Isopropylphenyl isocyanate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441570010

 View more versions of this product

Catalog No. AC441570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Isopropylphenyl isocyanate
88°C
Glass Bottle
66.0°C to 67.0°C (5.0mmHg)
1g
94.0
94% min. (GC)
1-isocyanato-2-isopropyl-benzene, 1-isocyanato-2-isopropylbenzene, 1-isocyanato-2-propan-2-yl benzene, 2-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate, o-isopropylphenyl isocyanate
CC(C)C1=CC=CC=C1N=C=O
161.21
161.21
95%
1.0190g/mL
Authentic
1.019
Colorless
56309-56-9
100.0
C10H11NO
MWBSQVPLYJZPTG-UHFFFAOYSA-N
1-isocyanato-2-propan-2-ylbenzene
91797
Liquid

Chemical Identifiers

56309-56-9
161.21
1-isocyanato-2-isopropyl-benzene, 1-isocyanato-2-isopropylbenzene, 1-isocyanato-2-propan-2-yl benzene, 2-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate, o-isopropylphenyl isocyanate
1-isocyanato-2-propan-2-ylbenzene
C10H11NO
MWBSQVPLYJZPTG-UHFFFAOYSA-N
91797
CC(C)C1=CC=CC=C1N=C=O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 260-105-4

Documents