Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropylbenzimidazole, 98%

$159.72 - $2,041.66

Chemical Identifiers

Chemical Name or Material 2-Isopropylbenzimidazole
CAS 5851-43-4
Molecular Formula C10H12N2
Formula Weight 160.22
MDL Number MFCD00051759
Beilstein 3535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1135606

  SDS
alfa aesar™
A1135606
5g Each for $159.72

AAA1135622

  SDS
alfa aesar™
A1135622
100g Each for $2,041.66

AAA1135614

  SDS
alfa aesar™
A1135614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Isopropylbenzimidazole
Melting Point 233° to 237°C
Quantity 5g
CAS 5851-43-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DD9014000
Assay 98%
Molecular Formula C10H12N2
Formula Weight 160.22
MDL Number MFCD00051759
Beilstein 3535
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.