missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropylanisole, 98%

Manufacturer:  Alfa Aesar™ H5338322

 View more versions of this product

Catalog No. AAH5338322



Description

Description

Applications
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Isopropylanisole
198°C to 200°C
2944-47-0
MFCD00798590
NNZRVXTXKISCGS-UHFFFAOYSA-N
1-methoxy-2-propan-2-ylbenzene
270623
98%
86°C (187°F)
100g
C10H14O
1-isopropyl-2-methoxybenzene, 2-isopropylanisole, o-isopropylanisole, anisole, o-isopropyl, benzene, 1-methoxy-2-1-methylethyl, 2-o-methoxyphenyl propane, 1-methoxy-2-propan-2-yl benzene, isopropylanisol, isopropyl anisole, acmc-20ansj
CC(C)C1=CC=CC=C1OC
150.221
150.22

Chemical Identifiers

2944-47-0
150.221
NNZRVXTXKISCGS-UHFFFAOYSA-N
270623
CC(C)C1=CC=CC=C1OC
C10H14O
MFCD00798590
1-isopropyl-2-methoxybenzene, 2-isopropylanisole, o-isopropylanisole, anisole, o-isopropyl, benzene, 1-methoxy-2-1-methylethyl, 2-o-methoxyphenyl propane, 1-methoxy-2-propan-2-yl benzene, isopropylanisol, isopropyl anisole, acmc-20ansj
1-methoxy-2-propan-2-ylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents