Please login to your online account to display your discounted pricing

2-Isopropylaniline, 97%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC259395000

 View more versions of this product

Catalog No. AC259395000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 135.21
Quantity 500mL
Color Clear pale yellow to orange or brown
Density 0.9550g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Isopropylaniline
Infrared Spectrum Authentic
Linear Formula (CH3)2CHC6H4NH2
Synonym o-Aminocumene
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Assay Percent Range 97%
Molecular Formula C9H13N
MDL Number MFCD00007720
CAS 643-28-7
Packaging Glass bottle
Health Hazard 1 Warning
Refractive Index 1.5470 to 1.5500 (20°C, 589nm)
Storage Information Product may darken on storage
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Isopropylaniline
CAS 643-28-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H13N
Linear Formula (CH3)2CHC6H4NH2
Formula Weight 135.21
MDL Number MFCD00007720
Synonym o-Aminocumene
Accessories

Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents