missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Isopropylaniline, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC259391000

 View more versions of this product

Catalog No. AC259391000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Liquid
Clear pale yellow to orange or brown
96.0
1.5470 to 1.5500 (20°C, 589nm)
643-28-7
C9H13N
o-Aminocumene
Glass bottle
97%
135.21
100mL
0.9550g/mL
100.0
Product may darken on storage
MFCD00007720
Authentic
(CH3)2CHC6H4NH2
2-Isopropylaniline

Chemical Identifiers

643-28-7
100.0
C9H13N
135.21
o-Aminocumene
≥96.0%
96.0
97%
(CH3)2CHC6H4NH2
MFCD00007720
2-Isopropylaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning