Learn More
2-Isopropylaniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 259390050
Specifications
2-Isopropylaniline | |
96.0 | |
0.9550g/mL | |
95°C | |
96% min. (GC) | |
C9H13N | |
(CH3)2CHC6H4NH2 | |
5mL | |
2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline | |
YKOLZVXSPGIIBJ-UHFFFAOYSA-N | |
2-propan-2-ylaniline | |
12561 | |
97% |
643-28-7 | |
100.0 | |
112.0°C to 113.0°C (18.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5470 to 1.5500 (20°C, 589nm) | |
MFCD00007720 | |
0.955 | |
Solubility in water: insoluble | |
CC(C)C1=CC=CC=C1N | |
135.21 | |
135.21 |
Chemical Identifiers
643-28-7 | |
135.21 | |
YKOLZVXSPGIIBJ-UHFFFAOYSA-N | |
12561 | |
CC(C)C1=CC=CC=C1N |
C9H13N | |
MFCD00007720 | |
2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline | |
2-propan-2-ylaniline |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 211-397-7
Recommended Storage : Product may darken on storage
RUO â Research Use Only