missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropylamino-2'-methoxyacetophenone

$78.04 - $78.04

Chemical Identifiers

Molecular Formula C12H17NO2
Formula Weight 207.27
MDL Number MFCD08064237
Chemical Name or Material 2-Isopropylamino-2'-methoxyacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50734MD SDS Alfa Aesar™
H50734MD
250mg Each for $78.04
AAH5073403 SDS Alfa Aesar™
H5073403
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C12H17NO2
207.27
MFCD08064237
2-Isopropylamino-2'-methoxyacetophenone

Specifications

250mg
207.27
2-Isopropylamino-2'-methoxyacetophenone
C12H17NO2
MFCD08064237