Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropylamino-1-phenylethanol

$112.51 - $389.43

Chemical Identifiers

Chemical Name or Material 2-Isopropylamino-1-phenylethanol
CAS 4164-21-0
Molecular Formula C11H17NO
Formula Weight 179.26
MDL Number MFCD01732724
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5079303

  SDS
alfa aesar™
H5079303
1g Each for $389.43

AAH50793MD

  SDS
alfa aesar™
H50793MD
250mg Each for $112.51
Description & Specifications

Specifications

Formula Weight 179.26
MDL Number MFCD01732724
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DO8500000
Quantity 1g
Chemical Name or Material 2-Isopropylamino-1-phenylethanol
Molecular Formula C11H17NO
CAS 4164-21-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.