missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropylamino-1-(3-methoxyphenyl)ethanol

$88.05 - $88.05

Chemical Identifiers

Molecular Formula C12H19NO2
Formula Weight 209.29
MDL Number MFCD08060963
Chemical Name or Material 2-Isopropylamino-1-(3-methoxyphenyl)ethanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5073703 SDS Alfa Aesar™
H5073703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH50737MD SDS Alfa Aesar™
H50737MD
250mg Each for $88.05
Specifications Description & Specifications

Chemical Identifiers

C12H19NO2
209.29
MFCD08060963
2-Isopropylamino-1-(3-methoxyphenyl)ethanol

Specifications

1g
209.29
2-Isopropylamino-1-(3-methoxyphenyl)ethanol
C12H19NO2
MFCD08060963

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning