Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropyl-6-methylphenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-Isopropyl-6-methylphenyl isocyanate
CAS 102561-43-3
Molecular Formula C11H13NO
Formula Weight 175.23
MDL Number MFCD00019911
UN Number UN2206
Beilstein 6368845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1270303

 
alfa aesar™
L1270303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1270306

 
alfa aesar™
L1270306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.23
Chemical Name or Material 2-Isopropyl-6-methylphenyl isocyanate
Molecular Formula C11H13NO
CAS 102561-43-3
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
MDL Number MFCD00019911
UN Number UN2206
Beilstein 6368845
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Flash Point 103°C (217°F)
Boiling Point 91° to 92°C (3mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C