Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropyl-5-methyl-2-hexenal, cis + trans, 96%

Chemical Identifiers

Chemical Name or Material 2-Isopropyl-5-methyl-2-hexenal
CAS 35158-25-9
Name Note Cis + Trans
Molecular Formula C10H18O
Formula Weight 154.25
MDL Number MFCD00036616
UN Number UN1989
Beilstein 1752384
Synonym Isodehydrolavandulal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0719309

 
alfa aesar™
L0719309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0719318

 
alfa aesar™
L0719318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.25
CAS 35158-25-9
Chemical Name or Material 2-Isopropyl-5-methyl-2-hexenal
Assay 96%
Molecular Formula C10H18O
MDL Number MFCD00036616
Beilstein 1752384
Synonym Isodehydrolavandulal
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 55°C (24mmHg)
TSCA TSCA
Name Note Cis + Trans
UN Number UN1989
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
RTECS Number MP6450000
Flash Point 60°C (140°F)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C