missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropoxypyridine-5-boronic acid, 97%

Manufacturer:  Alfa Aesar™ H6446106

 View more versions of this product

Catalog No. AAH6446106



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

5g
C8H12BNO3
2-isopropoxy-5-pyridineboronic acid, 2-isopropoxypyridine-5-boronic acid, 2-isoproxypyridine-5-boronic acid, 6-isopropoxy-3-pyridyl boronic acid, 6-isopropoxypyridin-3-yl boronic acid, 6-isopropoxypyridin-3-ylboronic acid, 6-isopropoxypyridine-3-boronic acid, 6-isoproxypyridine-3-boronic acid, 6-propan-2-yloxy pyridin-3-yl boronic acid, boronic acid, 6-1-methylethoxy-3-pyridinyl
SGEOBUJTURUUJE-UHFFFAOYSA-N
(6-propan-2-yloxypyridin-3-yl)boronic acid
44754864
Solid
870521-30-5
MFCD07368870
Slightly soluble in water.
B(C1=CN=C(C=C1)OC(C)C)(O)O
180.998
181
97%

Chemical Identifiers

870521-30-5
180.998
SGEOBUJTURUUJE-UHFFFAOYSA-N
44754864
B(C1=CN=C(C=C1)OC(C)C)(O)O
C8H12BNO3
MFCD07368870
2-isopropoxy-5-pyridineboronic acid, 2-isopropoxypyridine-5-boronic acid, 2-isoproxypyridine-5-boronic acid, 6-isopropoxy-3-pyridyl boronic acid, 6-isopropoxypyridin-3-yl boronic acid, 6-isopropoxypyridin-3-ylboronic acid, 6-isopropoxypyridine-3-boronic acid, 6-isoproxypyridine-3-boronic acid, 6-propan-2-yloxy pyridin-3-yl boronic acid, boronic acid, 6-1-methylethoxy-3-pyridinyl
(6-propan-2-yloxypyridin-3-yl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents