Please login to your online account to display your discounted pricing

2-Isopropoxyphenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Isopropoxyphenol
Name Note 97%
CAS 4812-20-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H12O2
Linear Formula (CH3)2CHOC6H4OH
Formula Weight 152.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC209220250

  SDS
thermo fisher scientific
209220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC209220050

  SDS
thermo fisher scientific
209220050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 152.19
Quantity 25g
Color Clear light yellow to light red-brown
EINECS Number 225-379-1
TSCA TSCA
Density 1.0300g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Isopropoxyphenol
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H12O2
Linear Formula (CH3)2CHOC6H4OH
MDL Number MFCD00002184
CAS 4812-20-8
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5140 to 1.5160 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.