Learn More
2-Isopropoxyphenol, 97%, Thermo Scientific Chemicals
$74.40 - $259.12
Chemical Identifiers
CAS | 4812-20-8 |
---|---|
Molecular Formula | C9H12O2 |
Molecular Weight (g/mol) | 152.193 |
MDL Number | MFCD00002184 |
InChI Key | ZNCUUYCDKVNVJH-UHFFFAOYSA-N |
Synonym | 2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a |
PubChem CID | 20949 |
ChEBI | CHEBI:38547 |
IUPAC Name | 2-propan-2-yloxyphenol |
SMILES | CC(C)OC1=CC=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0736006
|
Thermo Scientific Chemicals
L0736006 |
5 g |
Each for $74.40
|
|
|||||
AAL0736014
|
Thermo Scientific Chemicals
L0736014 |
25 g |
Each for $259.12
|
|
|||||
Description
2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
4812-20-8 | |
152.193 | |
ZNCUUYCDKVNVJH-UHFFFAOYSA-N | |
20949 | |
2-propan-2-yloxyphenol |
C9H12O2 | |
MFCD00002184 | |
2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a | |
CHEBI:38547 | |
CC(C)OC1=CC=CC=C1O |
Specifications
4812-20-8 | |
100°C to 102°C (11 mmHg) | |
C9H12O2 | |
MFCD00002184 | |
1937063 | |
Soluble in water. | |
CC(C)OC1=CC=CC=C1O | |
152.193 | |
CHEBI:38547 | |
97% |
1.032 | |
87°C (188°F) | |
1.515 | |
5 g | |
2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a | |
ZNCUUYCDKVNVJH-UHFFFAOYSA-N | |
2-propan-2-yloxyphenol | |
20949 | |
152.19 | |
2-Isopropoxyphenol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 225-379-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only