missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropoxybenzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H5316803

 View more versions of this product

Catalog No. AAH5316803



Specifications

Specifications

2-Isopropoxybenzeneboronic acid
199°C
1g
C9H13BO3
2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297
B(C1=CC=CC=C1OC(C)C)(O)O
180.01
180.01
110°C (230°F)
30°C to 35°C
138008-97-6
MFCD03427050
XDMKBIIRBDPSOE-UHFFFAOYSA-N
(2-propan-2-yloxyphenyl)boronic acid
5084099
95%

Chemical Identifiers

138008-97-6
180.01
XDMKBIIRBDPSOE-UHFFFAOYSA-N
5084099
B(C1=CC=CC=C1OC(C)C)(O)O
C9H13BO3
MFCD03427050
2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297
(2-propan-2-yloxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents