Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isopropoxybenzaldehyde, 98%

$1,228.35 - $1,228.35

Chemical Identifiers

Chemical Name or Material 2-Isopropoxybenzaldehyde
CAS 22921-58-0
Molecular Formula C10H12O2
Formula Weight 164.21
MDL Number MFCD01571834
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5048706

 
alfa aesar™
H5048706
5g Each for $1,228.35
Description & Specifications

Specifications

Formula Weight 164.21
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Chemical Name or Material 2-Isopropoxybenzaldehyde
Assay 98%
Molecular Formula C10H12O2
MDL Number MFCD01571834
CAS 22921-58-0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Isopropoxybenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.