missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
Supplier: TCI America I0658100G
Specifications
| 2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| Colorless | |
| C9H19BO3 | |
| 100 g | |
| 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane | |
| B1(OC(C(O1)(C)C)(C)C)OC(C)C | |
| 186.058 | |
| 186.06 | |
| Liquid |
| 61676-62-8 | |
| 80°C | |
| MFCD00192241 | |
| 1993 | |
| MRWWWZLJWNIEEJ-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane | |
| 5017741 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 61676-62-8 | |
| 186.058 | |
| MRWWWZLJWNIEEJ-UHFFFAOYSA-N | |
| 5017741 | |
| B1(OC(C(O1)(C)C)(C)C)OC(C)C |
| C9H19BO3 | |
| MFCD00192241 | |
| 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane | |
| 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No