missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%, Thermo Scientific™
$101.21 - $101.21
Chemical Identifiers
CAS | 61676-62-8 |
---|---|
Molecular Formula | C9H19BO3 |
Molecular Weight (g/mol) | 186.06 |
MDL Number | MFCD00192241 |
InChI Key | MRWWWZLJWNIEEJ-UHFFFAOYSA-N |
Synonym | 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane |
PubChem CID | 5017741 |
IUPAC Name | 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)OC(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC359490050
|
Thermo Scientific Chemicals
359490050 |
5 g | Glass bottle |
Each for $101.21
|
|
Chemical Identifiers
61676-62-8 | |
186.06 | |
MRWWWZLJWNIEEJ-UHFFFAOYSA-N | |
5017741 | |
B1(OC(C(O1)(C)C)(C)C)OC(C)C |
C9H19BO3 | |
MFCD00192241 | |
2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane | |
4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane |
Specifications
61676-62-8 | |
100.0 | |
0.9120g/mL | |
42°C | |
Authentic | |
MFCD00192241 | |
1.4080 to 1.41 | |
2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane | |
MRWWWZLJWNIEEJ-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane | |
5017741 | |
97% | |
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97% |
96.0 | |
Colorless | |
73°C (15.0mmHg) | |
96% min. (GC) | |
C9H19BO3 | |
Glass bottle | |
5 g | |
0.912 | |
B1(OC(C(O1)(C)C)(C)C)OC(C)C | |
186.06 | |
186.06 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and k
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
RUO â Research Use Only