missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC359490010

 View more versions of this product

Catalog No. AC359490010



Specifications

Specifications

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
42°C
Glass bottle
0.912
Colorless
97%
96.0
96% min. (GC)
MFCD00192241
MRWWWZLJWNIEEJ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane
5017741
Liquid
0.9120g/mL
Authentic
1.4080 to 1.41
73°C (15.0mmHg)
1g
61676-62-8
100.0
C9H19BO3
2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)OC(C)C
186.06
186.06
97%

Chemical Identifiers

61676-62-8
186.06
MRWWWZLJWNIEEJ-UHFFFAOYSA-N
5017741
B1(OC(C(O1)(C)C)(C)C)OC(C)C
C9H19BO3
MFCD00192241
2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and k

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS Documents