Please login to your online account to display your discounted pricing

2-Isonitrosoacetophenone 99%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  122630050

 View more versions of this product

Catalog No. AC122630050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 149.15
Quantity 5g
Color Beige to yellow
TSCA TSCA
RTECS Number MD3325000
Assay 98.5 to 101.5%
EINECS Number 208-539-5
Melting Point 122° to 126°C
Beilstein 07, 671
Merck Index 15, 5237
CAS Max % 100.0
Infrared Spectrum Authentic
Suitability suitable for determination of iron (Fe II)
CAS Min % 98.5
Synonym 2-Hydroxyiminoacetophenone
Assay Percent Range 99%
Molecular Formula C8H7NO2
Name Note 99%
CAS 532-54-7
Chemical Name or Material 2-Isonitrosoacetophenone

Specifications

Chemical Name or Material 2-Isonitrosoacetophenone
Name Note 99%
CAS 532-54-7
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 to 101.5%
Assay Percent Range 99%
Molecular Formula C8H7NO2
Formula Weight 149.15
Synonym 2-Hydroxyiminoacetophenone