missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Isonitrosoacetophenone 99%, ACROS Organics™

Chemical Identifiers

CAS 532-54-7
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8H7NO2
Formula Weight 149.15
Synonym 2-Hydroxyiminoacetophenone
Chemical Name or Material 2-Isonitrosoacetophenone, 99%
Assay 98.5 to 101.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC122630050 SDS Thermo Fisher Scientific
122630050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

532-54-7
100.0
C8H7NO2
2-Hydroxyiminoacetophenone
98.5 to 101.5%
98.5
99%
149.15
2-Isonitrosoacetophenone, 99%

Specifications

5g
98.5
Crystalline Powder
C8H7NO2
07, 671
2-Hydroxyiminoacetophenone
suitable for determination of iron (Fe II)
122° to 126°C
98.5 to 101.5%
532-54-7
100.0
99%
149.15
15, 5237
Authentic
Beige to yellow
2-Isonitrosoacetophenone, 99%

Safety and Handling

EINECSNumber : 208-539-5

RTECSNumber : MD3325000

TSCA : TSCA