missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isocyanatoethyl Acrylate (stabilized with BHT) 98.0+%, TCI America™
Supplier: TCI America I09185G
Specifications
| 2-Isocyanatoethyl Acrylate (stabilized with BHT) | |
| Colorless | |
| C6H7NO3 | |
| 5 g | |
| Acrylic Acid 2-Isocyanatoethyl Ester, 2-(Acryloyloxy)ethyl Isocyanate | |
| C=CC(=O)OCCN=C=O | |
| 141.126 | |
| 141.13 | |
| Liquid |
| 13641-96-8 | |
| 82°C | |
| MFCD11112219 | |
| 2206 | |
| DPNXHTDWGGVXID-UHFFFAOYSA-N | |
| 2-isocyanatoethyl prop-2-enoate | |
| 3014768 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 13641-96-8 | |
| 141.126 | |
| DPNXHTDWGGVXID-UHFFFAOYSA-N | |
| 3014768 | |
| C=CC(=O)OCCN=C=O |
| C6H7NO3 | |
| MFCD11112219 | |
| Acrylic Acid 2-Isocyanatoethyl Ester, 2-(Acryloyloxy)ethyl Isocyanate | |
| 2-isocyanatoethyl prop-2-enoate |
Safety and Handling
EINECSNumber : (2)-4063
TSCA : No
Recommended Storage : Refrigerator