Please login to your online account to display your discounted pricing

2-Isocyanatobenzonitrile, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB13667DA

 View more versions of this product

Catalog No. BTB13667DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 144.13
Quantity 1g
Assay 95%
Molecular Formula C8H4N2O
Chemical Name or Material 2-Isocyanatobenzonitrile
Assay Percent Range 95%
CAS 42066-86-4
Packaging Amber glass bottle
Linear Formula CNC6H4NCO

Specifications

Chemical Name or Material 2-Isocyanatobenzonitrile
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H4N2O
Linear Formula CNC6H4NCO
Formula Weight 144.13