Learn More
2-Isobutylthiazole, 99%, Thermo Scientific Chemicals
$97.42 - $333.97
Chemical Identifiers
CAS | 18640-74-9 |
---|---|
Molecular Formula | C7H11NS |
Molecular Weight (g/mol) | 141.232 |
MDL Number | MFCD00005334 |
InChI Key | CMPVUVUNJQERIT-UHFFFAOYSA-N |
Synonym | 2-isobutylthiazole, 2-isobutyl-1,3-thiazole, 2-2-methylpropyl thiazole, thiazole, 2-isobutyl, 2-2-methylpropyl-1,3-thiazole, 2-isobutyl thiazole, thiazole, 2-2-methylpropyl, iso-butyl thiazole, fema no. 3134 |
PubChem CID | 62725 |
IUPAC Name | 2-(2-methylpropyl)-1,3-thiazole |
SMILES | CC(C)CC1=NC=CS1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1375906
|
Thermo Scientific Chemicals
A1375906 |
5 g |
Each for $97.42
|
|
|||||
AAA1375914
|
Thermo Scientific Chemicals
A1375914 |
25 g |
Each for $333.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18640-74-9 | |
141.232 | |
CMPVUVUNJQERIT-UHFFFAOYSA-N | |
62725 | |
CC(C)CC1=NC=CS1 |
C7H11NS | |
MFCD00005334 | |
2-isobutylthiazole, 2-isobutyl-1,3-thiazole, 2-2-methylpropyl thiazole, thiazole, 2-isobutyl, 2-2-methylpropyl-1,3-thiazole, 2-isobutyl thiazole, thiazole, 2-2-methylpropyl, iso-butyl thiazole, fema no. 3134 | |
2-(2-methylpropyl)-1,3-thiazole |
Specifications
18640-74-9 | |
172°C to 173°C | |
Characteristic | |
1.496 | |
5 g | |
507823 | |
CMPVUVUNJQERIT-UHFFFAOYSA-N | |
2-(2-methylpropyl)-1,3-thiazole | |
62725 | |
99% |
0.998 | |
58°C (136°F) | |
C7H11NS | |
MFCD00005334 | |
UN1993 | |
2-isobutylthiazole, 2-isobutyl-1,3-thiazole, 2-2-methylpropyl thiazole, thiazole, 2-isobutyl, 2-2-methylpropyl-1,3-thiazole, 2-isobutyl thiazole, thiazole, 2-2-methylpropyl, iso-butyl thiazole, fema no. 3134 | |
CC(C)CC1=NC=CS1 | |
141.232 | |
141.24 | |
2-Isobutylthiazole |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 242-470-1
RTECSNumber : XJ5103412
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only