Learn More
2-Isobutylpyridine, 99%, Thermo Scientific Chemicals
$134.74 - $134.74
Chemical Identifiers
CAS | 6304-24-1 |
---|---|
Molecular Formula | C9H13N |
Molecular Weight (g/mol) | 135.21 |
MDL Number | MFCD02262142 |
InChI Key | BBVSPSDWPYWMOR-UHFFFAOYSA-N |
Synonym | 2-isobutylpyridine, 2-2-methylpropyl pyridine, 2-isobutyl pyridine, pyridine, 2-2-methylpropyl, unii-gs3k23h1q8, pyridine, 2-isobutyl, fema no. 3370, 2-isobutyl-pyridin, 2-isobutyl-pyridine, acmc-1b4hh |
PubChem CID | 61385 |
IUPAC Name | 2-(2-methylpropyl)pyridine |
SMILES | CC(C)CC1=CC=CC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2234906
|
Thermo Scientific Chemicals
B2234906 |
5 g |
Each for $134.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6304-24-1 | |
135.21 | |
BBVSPSDWPYWMOR-UHFFFAOYSA-N | |
61385 | |
CC(C)CC1=CC=CC=N1 |
C9H13N | |
MFCD02262142 | |
2-isobutylpyridine, 2-2-methylpropyl pyridine, 2-isobutyl pyridine, pyridine, 2-2-methylpropyl, unii-gs3k23h1q8, pyridine, 2-isobutyl, fema no. 3370, 2-isobutyl-pyridin, 2-isobutyl-pyridine, acmc-1b4hh | |
2-(2-methylpropyl)pyridine |
Specifications
6304-24-1 | |
C9H13N | |
5 g | |
BBVSPSDWPYWMOR-UHFFFAOYSA-N | |
2-(2-methylpropyl)pyridine | |
61385 | |
99% |
0.899 | |
MFCD02262142 | |
2-isobutylpyridine, 2-2-methylpropyl pyridine, 2-isobutyl pyridine, pyridine, 2-2-methylpropyl, unii-gs3k23h1q8, pyridine, 2-isobutyl, fema no. 3370, 2-isobutyl-pyridin, 2-isobutyl-pyridine, acmc-1b4hh | |
CC(C)CC1=CC=CC=N1 | |
135.21 | |
135.21 | |
2-Isobutylpyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
EINECSNumber : 228-606-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only