Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isobutylpyrazine, 98%

$67.58 - $823.51

Chemical Identifiers

Chemical Name or Material 2-Isobutylpyrazine
CAS 29460-92-2
Molecular Formula C8H12N2
Formula Weight 136.2
MDL Number MFCD00156150
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1024503

 
alfa aesar™
A1024503
1g Each for $67.58

AAA1024514

 
alfa aesar™
A1024514
25g Each for $823.51
Description & Specifications

Specifications

Formula Weight 136.2
Chemical Name or Material 2-Isobutylpyrazine
Assay 98%
Molecular Formula C8H12N2
MDL Number MFCD00156150
CAS 29460-92-2
Quantity 1g