Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isobutyl-4,5-dimethylthiazole, 99%

$75.03 - $75.03

Chemical Identifiers

Chemical Name or Material 2-Isobutyl-4,5-dimethylthiazole
CAS 53498-32-1
Molecular Formula C9H15NS
Formula Weight 169.28
MDL Number MFCD00053136
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2255409

  SDS
alfa aesar™
B2255409
10g Each for $75.03
Description & Specifications

Specifications

Formula Weight 169.28
Molecular Formula C9H15NS
Quantity 10g
Assay 99%
MDL Number MFCD00053136
Chemical Name or Material 2-Isobutyl-4,5-dimethylthiazole
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
CAS 53498-32-1
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.