Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isobutyl-4,5-dimethyl-3-thiazoline, 97%

$34.49 - $124.05

Chemical Identifiers

Chemical Name or Material 2-Isobutyl-4,5-dimethyl-3-thiazoline
CAS 65894-83-9
Molecular Formula C9H17NS
Formula Weight 171.3
MDL Number MFCD01680822
Synonym 2-(2-Methylpropyl)-4,5-dimethyl-3-thiazoline; 4,5-Dimethyl-2-isobutyl-3-thiazoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2637903

 
alfa aesar™
H2637903
1g Each for $34.49

AAH2637909

 
alfa aesar™
H2637909
10g Each for $124.05
Description & Specifications

Specifications

Formula Weight 171.3
Chemical Name or Material 2-Isobutyl-4,5-dimethyl-3-thiazoline
TSCA TSCA
Assay 97%
Molecular Formula C9H17NS
MDL Number MFCD01680822
CAS 65894-83-9
Synonym 2-(2-Methylpropyl)-4,5-dimethyl-3-thiazoline; 4,5-Dimethyl-2-isobutyl-3-thiazoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 208°C
Flash Point 87°C (189°F)
RTECS Number XJ6642800
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.