Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Isobutyl-1,3-dithiane, 97%

$35.55 - $414.36

Chemical Identifiers

Chemical Name or Material 2-Isobutyl-1,3-dithiane
CAS 69824-23-3
Molecular Formula C8H16S2
Formula Weight 176.35
MDL Number MFCD00040290
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2147122

  SDS
alfa aesar™
B2147122
100g Each for $414.36

AAB2147106

  SDS
alfa aesar™
B2147106
5g Each for $35.55

AAB2147114

  SDS
alfa aesar™
B2147114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Isobutyl-1,3-dithiane
Quantity 100g
CAS 69824-23-3
Assay 97%
Molecular Formula C8H16S2
Formula Weight 176.35
MDL Number MFCD00040290