missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Isobutoxynaphthalene, TCI America™

$76.39 - $550.02

Chemical Identifiers

CAS 2173-57-1
Molecular Formula C14H16O
Molecular Weight (g/mol) 200.281
MDL Number MFCD00046467
InChI Key XOHIHZHSDMWWMS-UHFFFAOYSA-N
Synonym Isobutyl 2-Naphthyl Ether
PubChem CID 16582
IUPAC Name 2-(2-methylpropoxy)naphthalene
SMILES CC(C)COC1=CC2=CC=CC=C2C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0433500G View Documents TCI America
N0433500G
500g Each for $550.02
 
N043325G View Documents TCI America
N043325G
25g Each for $76.39
 
Specifications

Chemical Identifiers

2173-57-1
200.281
XOHIHZHSDMWWMS-UHFFFAOYSA-N
16582
CC(C)COC1=CC2=CC=CC=C2C=C1
C14H16O
MFCD00046467
Isobutyl 2-Naphthyl Ether
2-(2-methylpropoxy)naphthalene

Specifications

White-Yellow
2173-57-1
MFCD00046467
XOHIHZHSDMWWMS-UHFFFAOYSA-N
2-(2-methylpropoxy)naphthalene
16582
Crystal-Lump at 20°C
2-Isobutoxynaphthalene
500g
C14H16O
Isobutyl 2-Naphthyl Ether
CC(C)COC1=CC2=CC=CC=C2C=C1
200.281
200.28
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0362

RTECSNumber : KO1255000

TSCA : Yes