Learn More
2-Iodoxybenzoic acid, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 374650500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagentSpecifications
2-Iodoxybenzoic acid | |
250.0°C | |
Glass bottle | |
MFCD00024166 | |
15, 5091 | |
280.01 | |
39 to 50% as 2-Iodoxybenzoic acid |
61717-82-6,64297-64-9,121-91-5,65-85-0 | |
White | |
C7H5IO4 | |
50 g | |
Solubility in water: slightly soluble | |
280.01 | |
Powder, Prills or Agglomerates |
Chemical Identifiers
61717-82-6 | |
C7H5IO4 | |
280.01 | |
MFCD00024166 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only