missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 1228778-59-3 |
---|---|
Molecular Formula | C18H11I |
Molecular Weight (g/mol) | 354.19 |
InChI Key | ZMZNYRZSJDYTIO-UHFFFAOYSA-N |
PubChem CID | 88955426 |
IUPAC Name | 2-iodotriphenylene |
SMILES | C1=CC=C2C(=C1)C3=C(C=C(C=C3)I)C4=CC=CC=C24 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I0978200MG |
TCI America
I0978200MG |
200mg |
Each for $264.31
|
|
I09781G |
TCI America
I09781G |
1g |
Each for $913.63
|
|
Chemical Identifiers
1228778-59-3 | |
354.19 | |
88955426 | |
C1=CC=C2C(=C1)C3=C(C=C(C=C3)I)C4=CC=CC=C24 |
C18H11I | |
ZMZNYRZSJDYTIO-UHFFFAOYSA-N | |
2-iodotriphenylene |
Specifications
White-Yellow | |
200mg | |
C18H11I | |
C1=CC=C2C(=C1)C3=C(C=C(C=C3)I)C4=CC=CC=C24 | |
354.19 | |
354.19 | |
≥98.0% (GC) |
157°C | |
1228778-59-3 | |
ZMZNYRZSJDYTIO-UHFFFAOYSA-N | |
2-iodotriphenylene | |
88955426 | |
Crystal-Powder at 20°C | |
2-Iodotriphenylene |
Safety and Handling
TSCA : No