Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodothioanisole, 96%

$93.27 - $393.27

Chemical Identifiers

Chemical Name or Material 2-Iodothioanisole
CAS 33775-94-9
Molecular Formula C7H7IS
Formula Weight 250.1
MDL Number MFCD00051611
Beilstein 2516385
Synonym 2-Iodophenyl methyl sulfide; 2-(Methylthio)iodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1405106

 
alfa aesar™
A1405106
5g Each for $93.27

AAA1405114

 
alfa aesar™
A1405114
25g Each for $393.27
Description & Specifications

Specifications

Formula Weight 250.1
Assay 96%
Synonym 2-Iodophenyl methyl sulfide; 2-(Methylthio)iodobenzene
Boiling Point 129° to 131°C (3mmHg)
Quantity 5g
CAS 33775-94-9
MDL Number MFCD00051611
Beilstein 2516385
Molecular Formula C7H7IS
Chemical Name or Material 2-Iodothioanisole