Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Iodoresorcinol, 97%

$177.80 - $177.80

Chemical Identifiers

Chemical Name or Material 2-Iodoresorcinol
CAS 41046-67-7
Molecular Formula C6H5IO2
Formula Weight 236.01
MDL Number MFCD09701440
Synonym 2-Iodo-1,3-dihydroxybenzene; 2-Iodobenzene-1,3-diol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3594103

  SDS
alfa aesar™
H3594103
1g Each for $177.80
Description & Specifications

Specifications

Formula Weight 236.01
Synonym 2-Iodo-1,3-dihydroxybenzene; 2-Iodobenzene-1,3-diol
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C6H5IO2
Chemical Name or Material 2-Iodoresorcinol
Assay 97%
MDL Number MFCD09701440
CAS 41046-67-7
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Melting Point 107° to 109°C
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.