Learn More
2-Iodopyridine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 379330010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Iodopyridine | |
97.5 | |
1.9280g/mL | |
97.5% min. (GC) | |
C5H4IN | |
1g | |
2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou | |
IC1=CC=CC=N1 | |
205.00 | |
204.99 |
5029-67-4 | |
100.0 | |
52.0°C (0.9 mmHg) | |
Glass bottle | |
MFCD00464928 | |
1.928 | |
CCZWSTFVHJPCEM-UHFFFAOYSA-N | |
2-iodopyridine | |
221126 | |
98% |
Chemical Identifiers
5029-67-4 | |
205.00 | |
CCZWSTFVHJPCEM-UHFFFAOYSA-N | |
221126 | |
IC1=CC=CC=N1 |
C5H4IN | |
MFCD00464928 | |
2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou | |
2-iodopyridine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Warning
RUO â Research Use Only