Learn More
2-Iodophenylacetonitrile, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1276706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Iodophenylacetonitrile | |
1.75 | |
>110°C (230°F) | |
1.617 | |
5 g | |
2083106 | |
2-iodophenylacetonitrile, 2-2-iodophenyl acetonitrile, 2-iodobenzyl cyanide, o-iodophenylacetonitrile, benzeneacetonitrile, 2-iodo, o-iodobenzyl cyanide, acmc-20a7xu, 2-iodo-phenylacetonitrile, 2-iodo-phenyl-acetonitrile, 2-2-iodophenyl ethanenitrile | |
C1=CC=C(C(=C1)CC#N)I | |
243.047 | |
243.05 |
40400-15-5 | |
139°C to 140°C (13 mmHg) | |
C8H6IN | |
MFCD00040888 | |
UN3276 | |
Light sensitive | |
FPSGTRJUQLYLHE-UHFFFAOYSA-N | |
2-(2-iodophenyl)acetonitrile | |
4402282 | |
96% |
Chemical Identifiers
40400-15-5 | |
243.047 | |
FPSGTRJUQLYLHE-UHFFFAOYSA-N | |
4402282 | |
C1=CC=C(C(=C1)CC#N)I |
C8H6IN | |
MFCD00040888 | |
2-iodophenylacetonitrile, 2-2-iodophenyl acetonitrile, 2-iodobenzyl cyanide, o-iodophenylacetonitrile, benzeneacetonitrile, 2-iodo, o-iodobenzyl cyanide, acmc-20a7xu, 2-iodo-phenylacetonitrile, 2-iodo-phenyl-acetonitrile, 2-2-iodophenyl ethanenitrile | |
2-(2-iodophenyl)acetonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only