missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Iodophenylacetic acid +97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC394040250

 View more versions of this product

Catalog No. AC394040250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Iodophenylacetic acid
97.0
Authentic
Powder
97+%
IC6H4CH2CO2H
MFCD00046546
112° to 116°C
18698-96-9
100.0
Glass bottle
≥97.0%
C8H7IO2
262.04
Cream
25g

Chemical Identifiers

18698-96-9
100.0
C8H7IO2
262.04
2-Iodophenylacetic acid
97.0
97+%
IC6H4CH2CO2H
MFCD00046546
≥97.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.