Please login to your online account to display your discounted pricing

2-Iodomelatonin, Tocris


High affinity melatonin agonist

Quantity 10mg

Manufacturer: r & d systems  073710

 View more versions of this product

Catalog No. 073710

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 358.18
Chemical Name or Material 2-Iodomelatonin
CAS 93515-00-5
Synonym N-[2-(2-Iodo-5-methoxyindol-3-yl)ethyl]acetamide
Molecular Formula C13H15IN2O2
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material 2-Iodomelatonin
Assay Percent Range >98%
Molecular Formula C13H15IN2O2
Formula Weight 358.18
Synonym N-[2-(2-Iodo-5-methoxyindol-3-yl)ethyl]acetamide
Accessories

Desiccate at +4°C