missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Iodoethyl)benzene, 97%, stabilized with copper, Thermo Scientific Chemicals
$550.87 - $550.87
Chemical Identifiers
| CAS | 17376-04-4 |
|---|---|
| Molecular Formula | C8H9I |
| Molecular Weight (g/mol) | 232.07 |
| MDL Number | MFCD00039408 |
| InChI Key | KVTHPKXDLVYNCH-UHFFFAOYSA-N |
| Synonym | 2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene |
| PubChem CID | 28503 |
| IUPAC Name | 2-iodoethylbenzene |
| SMILES | C1=CC=C(C=C1)CCI |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC305250250
|
Thermo Scientific Chemicals
305250250 |
25 mL | Glass bottle |
Each for $550.87
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 17376-04-4 | |
| 232.07 | |
| KVTHPKXDLVYNCH-UHFFFAOYSA-N | |
| 28503 | |
| C1=CC=C(C=C1)CCI |
| C8H9I | |
| MFCD00039408 | |
| 2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene | |
| 2-iodoethylbenzene |
Specifications
| 17376-04-4 | |
| 100.0 | |
| 1.6000g/mL | |
| 110°C | |
| 96% min. (GC) | |
| C8H9I | |
| C6H5CH2CH2I | |
| 25 mL | |
| 2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene | |
| KVTHPKXDLVYNCH-UHFFFAOYSA-N | |
| 2-iodoethylbenzene | |
| 28503 | |
| 97% | |
| (2-Iodoethyl)benzene, 97% |
| 96.0 | |
| Brown-Red | |
| 122.0°C to 123.0°C (13.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.6000 to 1.6020 | |
| MFCD00039408 | |
| 1.6 | |
| Solubility in water: insoluble in water | |
| C1=CC=C(C=C1)CCI | |
| 232.07 | |
| 232.07 | |
| Liquid |
Safety and Handling
TSCA : TSCA
RUO â Research Use Only