missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Iodoethyl)benzene, 97%, stabilized with copper, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 305250050

 View more versions of this product

Catalog No. AC305250050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(2-Iodoethyl)benzene
17376-04-4
100.0
Glass bottle
Liquid
97%
C8H9I
232.07
Clear red-brown
97%
96.0
Authentic
1.6000 to 1.6020 (20°C, 589nm)
≥96.0%
1.6g/mL
C6H5CHCHI
MFCD00039408
5mL

Chemical Identifiers

17376-04-4
100.0
C8H9I
232.07
(2-Iodoethyl)benzene, 97%
96.0
97%
C6H5CHCHI
MFCD00039408
≥96.0%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA